missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propionic acid, 98%

Product Code. 11303416
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303416 50 g 50g
11393406 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303416 Supplier Thermo Scientific Alfa Aesar Supplier No. L12861.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 646-01-5 | C4H8O2S | 120.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-01-5
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00059635
InChI Key CAOMCZAIALVUPA-UHFFFAOYSA-N
Synonym 3-methylthio propionic acid, 3-methylthiopropionate, 3-methylthiopropionic acid, 4-thiapentanoic acid, 3-methylsulfanyl propanoic acid, propanoic acid, 3-methylthio, 3-methylmercaptopropionic acid, unii-f523ali47d, 3-methylthio propanoic acid, propionic acid, 3-methylthio
PubChem CID 563
ChEBI CHEBI:1438
IUPAC Name 3-methylsulfanylpropanoic acid
SMILES CSCCC(=O)O

Specifications

Melting Point ∼16°C
Density 1.16
Boiling Point 244°C to 247°C
Flash Point 104°C (219°F)
Odor Unpleasant
Linear Formula CH3S(CH2)2CO2H
Refractive Index 1.4885
Quantity 50 g
UN Number UN3265
Beilstein 1743054
Formula Weight 120.17
Percent Purity 98%
Chemical Name or Material 3-(Methylthio)propionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.