missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propionaldehyde, 97%

Product Code. 11462487
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462487 250 g 250g
11447846 10 g 10g
11452487 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462487 Supplier Thermo Scientific Alfa Aesar Supplier No. A14774.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3268-49-3 | C4H8OS | 104.167 g/mol

3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

Solubility
It is readily soluble in alcohol solvents such as ethanol, propylene and glycol oil. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3268-49-3
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00007022
InChI Key CLUWOWRTHNNBBU-UHFFFAOYSA-N
Synonym methional, 3-methylthio propionaldehyde, 3-methylthio propanal, propanal, 3-methylthio, 4-thiapentanal, 3-methylsulfanyl propanal, 3-methylmercapto propionaldehyde, 3-methylthiopropanal, propionaldehyde, 3-methylthio, 3-methylmercaptopropyl aldehyde
PubChem CID 18635
ChEBI CHEBI:49017
IUPAC Name 3-methylsulfanylpropanal
SMILES CSCCC=O

Specifications

Melting Point -58°C
Density 1.052
Boiling Point 165°C to 166°C
Flash Point 64°C (147°F)
Odor Strong
Linear Formula CH3S(CH2)2CHO
Refractive Index 1.4825
Quantity 250 g
UN Number UN2785
Beilstein 1739289
Sensitivity Air sensitive
Solubility Information It is readily soluble in alcohol solvents such as ethanol,propylene and glycol oil. It is insoluble in water.
Formula Weight 104.17
Percent Purity 98%
Chemical Name or Material 3-(Methylthio)propionaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.