missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propionaldehyde 95%, ACROS Organics™

Product Code. 10463330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10463330 5 g Glass bottle 5g
10452971 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10463330 Supplier Thermo Scientific Acros Supplier No. 414920050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3268-49-3
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
InChI Key CLUWOWRTHNNBBU-UHFFFAOYSA-N
Synonym methional, 3-methylthio propionaldehyde, 3-methylthio propanal, propanal, 3-methylthio, 4-thiapentanal, 3-methylsulfanyl propanal, 3-methylmercapto propionaldehyde, 3-methylthiopropanal, propionaldehyde, 3-methylthio, 3-methylmercaptopropyl aldehyde
PubChem CID 18635
ChEBI CHEBI:49017
IUPAC Name 3-methylsulfanylpropanal
SMILES CSCCC=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.05g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3SCH2CH2CHO
Quantity 5 g
Formula Weight 104.17
Percent Purity ≥96%
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)propionaldehyde, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.