missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propan-1-ol, Dr. Ehrenstorfer

Product Code. 30327325
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327325 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327325 Supplier Dr. Ehrenstorfer Supplier No. DREC15144230

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Methylthio)-1-propanol
CAS 505-10-2
Analyte or Component Names 3-(Methylthio)-1-propanol
Shipping Condition +20°C
Molecular Formula C4 H10 O S
Certifications/Compliance ISO/IEC 17025
Synonym 3-(Methylthio)-1-propanol, 3-(Methylsulfanyl)-1-propanol, 3-(Methylthio)propanol, 3-Hydroxypropyl methyl sulfide, 3-Methylmercapto-1-propanol, 4-Thiapentan-1-ol, Methionol, NSC 2859, γ-Methylmercaptopropyl alcohol
InChI Formula InChI=1S/C4H10OS/c1-6-4-2-3-5/h5H,2-4H2,1H3
SMILES CSCCCO
IUPAC Name 3-methylsulfanylpropan-1-ol
Molecular Weight (g/mol) 106.19
Quantity 1 mL
Formula Weight 106.0452 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.