missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)butyraldehyde, 95%

Product Code. 11479517
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479517 50 g 50g
11469517 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479517 Supplier Thermo Scientific Alfa Aesar Supplier No. B22033.18
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16630-52-7 | C5H10OS | 118.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16630-52-7
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.19
MDL Number MFCD00036622
InChI Key NCBDFIPMWRKPDU-UHFFFAOYNA-N
Synonym 3-methylthio butanal, 3-methylthio butyraldehyde, butanal, 3-methylthio, 3-methylsulfanyl butanal, beta-methylthio butyraldehyde, fema no. 3374, butyraldehyde, 3-methylthio, potato butyraldehyde, 3-methylthiobutyraldehyde, butanal,3-methylthio
PubChem CID 61845
IUPAC Name 3-methylsulfanylbutanal
SMILES CSC(C)CC=O

Specifications

Boiling Point 63°C to 65°C (11 mmHg)
Flash Point 62°C (144°F)
Quantity 50 g
Sensitivity Air Sensitive
Formula Weight 118.2
Percent Purity 95%
Chemical Name or Material 3-(Methylthio)butyraldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.