missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)benzoic acid, 97%

Product Code. p-7029707
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11312179

Brand: Thermo Scientific Alfa Aesar H27200.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 825-99-0 | C8H8O2S | 168.21 g/mol

It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 825-99-0
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00156993
InChI Key PZGADOOBMVLBJE-UHFFFAOYSA-N
Synonym 3-methylthio benzoic acid, 3-methylsulfanyl benzoic acid, benzoic acid, 3-methylthio, benzoic acid, m-methylthio, 3-mercaptobenzoic acid s-methyl ether, 3-methylsulfanyl-benzoic acid, 3-carboxythioanisole, pubchem10679
PubChem CID 220329
IUPAC Name 3-methylsulfanylbenzoic acid
SMILES CSC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 126°C to 132°C
Quantity 1 g
Formula Weight 168.21
Percent Purity 97%
Chemical Name or Material 3-(Methylthio)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.