missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)-1-propanol, TRC

Product Code. 30470385
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470385 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470385 Supplier Toronto Research Chemicals Supplier No. TRCM33077125G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Methylthio)-1-propanol
CAS 505-10-2
Molecular Formula C4 H10 O S
Quantity 25 g
Synonym 3-(Methylthio)-1-propanol, 3-(Methylsulfanyl)-1-propanol, 3-(Methylthio)propanol, 3-Hydroxypropyl methyl sulfide, 3-Methylmercapto-1-propanol, 4-Thiapentan-1-ol, Methionol, NSC 2859, γ-Methylmercaptopropyl alcohol
InChI Formula InChI=1S/C4H10OS/c1-6-4-2-3-5/h5H,2-4H2,1H3
SMILES CSCCCO
IUPAC Name 3-methylsulfanylpropan-1-ol
Molecular Weight (g/mol) 106.19
Formula Weight 106.0452
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.