missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsulphonylbenzoic acid, 95%, Thermo Scientific™

Product Code. 10667961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10667961 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10667961 Supplier Thermo Scientific Acros Supplier No. 424860025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5345-27-7
CAS Min % 94.0
CAS Max % 100.0
Melting Point 230°C to 238°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H8O4S
Linear Formula SO2CH3C6H4COOH
MDL Number MFCD00216488
Quantity 2.5 g
Synonym 3-methylsulfonyl benzoic acid, 3-methylsulphonyl benzoic acid, 3-methylsulphonylbenzoic acid, 3-methanesulfonylbenzoic acid, 3-methanesulfonyl-benzoic acid, benzoic acid, 3-methylsulfonyl, 3-mesylbenzoic acid, pubchem10870, acmc-1auq1
InChI Key KUTBMATZUQWFSR-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=CC(=C1)C(=O)O
IUPAC Name 3-methylsulfonylbenzoic acid
Molecular Weight (g/mol) 200.22
PubChem CID 220380
Formula Weight 200.22
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Methylsulphonylbenzoic acid, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.