missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylsulfonylaminomethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15420046
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420046 1 g 1g
15430046 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420046 Supplier Thermo Scientific Alfa Aesar Supplier No. H52705.03
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 850568-39-7 | Molecular Formula: C8H12BNO4S | Molecular Weight: 229.057 g/mol

(3-Methylsulfonylaminomethyl)benzeneboronic acid, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Methylsulfonylaminomethyl)benzeneboronic acid, is used as a pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-39-7
Molecular Formula C8H12BNO4S
Molecular Weight (g/mol) 229.057
MDL Number MFCD06659817
InChI Key FJSWJJOLFPIDER-UHFFFAOYSA-N
Synonym 3-methylsulfonamidomethyl phenyl boronic acid, 3-methylsulphonylamino methyl benzeneboronic acid, 3-methanesulfonylaminomethylphenyl boronic acid, 3-methanesulfonylaminomethyl phenylboronic acid, 3-methanesulfonylaminomethyl benzeneboronic acid, 3-methanesulfonamidomethyl phenylboronic acid, 3-methylsulfonylaminomethyl benzeneboronic acid, 3-methylsulfonamidomethyl phenylboronic acid, 3-methylsulfonamidomethyl phenyl boronicacid, 3-methanesulfonamidomethyl phenyl boronic acid
PubChem CID 22309453
IUPAC Name [3-(methanesulfonamidomethyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)CNS(=O)(=O)C)(O)O

Specifications

Melting Point 246°C to 248°C
Quantity 1 g
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 229.06
Percent Purity 98%
Chemical Name or Material (3-Methylsulfonylaminomethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.