missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylsulfonylamino)benzeneboronic acid, 97%

Product Code. 15409677
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409677 1 g 1g
15419677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409677 Supplier Thermo Scientific Alfa Aesar Supplier No. H52895.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 148355-75-3 | C7H10BNO4S | 215.03 g/mol

Benzeneboronic acid mediates the ortho specific alpha hydroxyalkylation of phenols by aldehydes. The chemical itself can be used as a template for Diels-Alder reaction by forming linkage with a hydroxy diene and a hydroxy dieneophile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzeneboronic acid mediates the ortho specific alpha hydroxyalkylation of phenols by aldehydes. The chemical itself can be used as a template for Diels-Alder reaction by forming linkage with a hydroxy diene and a hydroxy dieneophile.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Keep Cold
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148355-75-3
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.03
MDL Number MFCD02179478
InChI Key XUIQQIRLFMCWLN-UHFFFAOYSA-N
Synonym 3-methylsulfonylamino phenylboronic acid, 3-methylsulfonamido phenylboronic acid, 3-methanesulfonylaminophenylboronic acid, 3-methylsulfonylaminophenylboronic acid, 3-methanesulfonamidophenyl boronic acid, 3-methanesulfonamido phenyl boronic acid, 3-methanesulfonamidophenylboronic acid, 3-methylsulphonylamino benzeneboronic acid, n-3-methanesulfonamidephenylboronic acid, 3-methylsulfonylamino benzeneboronic acid
PubChem CID 2773535
IUPAC Name [3-(methanesulfonamido)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)NS(=O)(=O)C)(O)O

Specifications

Melting Point 90°C to 96°C
Quantity 1 g
Formula Weight 215.03
Percent Purity 97%
Chemical Name or Material 3-(Methylsulfonylamino)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.