missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyridazine, 99%

Product Code. 10347170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347170 10 g Glass bottle 10g
10686342 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347170 Supplier Thermo Scientific Acros Supplier No. 148080100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1632-76-4 | C5H6N2 | 94.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1632-76-4
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006469
InChI Key MXDRPNGTQDRKQM-UHFFFAOYSA-N
Synonym pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j
PubChem CID 74208
IUPAC Name 3-methylpyridazine
SMILES CC1=NN=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.0310g/mL
Boiling Point 214°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5140 to 1.516
Quantity 10 g
Beilstein 23, 92
Specific Gravity 1.031
Formula Weight 94.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methylpyridazine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylpyridazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.