missing translation for 'onlineSavingsMsg'
Learn More

3-methylpyrazole-4-boronic acid, 97%, Thermo Scientific™

Product Code. 10235914
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10235914 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10235914 Supplier Thermo Scientific Maybridge Supplier No. MAY00037DA
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 936250-20-3
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H17BN2O2
MDL Number MFCD08690235
Quantity 1 g
Synonym 3-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 3-methyl-1h-pyrazole-4-boronic acid pinacol ester, 5-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 3-methylpyrazole-4-boronic acid, pinacol ester, 3-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 3-methylpyrazole-4-boronic acid pinacol ester, 3-methyl-4-pyrazole boronic acid pinacol ester, 3-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 3-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyrazole, 1h-pyrazole, 3-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
InChI Key MSJAEFFWTBMIKT-UHFFFAOYSA-N
SMILES CC1=C(C=NN1)B1OC(C)(C)C(C)(C)O1
IUPAC Name 5-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular Weight (g/mol) 208.07
PubChem CID 17750270
Formula Weight 208.07
Percent Purity 97%
Chemical Name or Material 3-Methylpyrazole-4-boronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.