missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpiperidine, 97%

Product Code. 11363037
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363037 100 g 100g
11353037 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363037 Supplier Thermo Scientific Alfa Aesar Supplier No. L13408.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626-56-2 | C6H13N | 99.177 g/mol

3-Methylpiperidine, is used as a reactant for synthesis of Phenylpropenamide derivatives for anti-hepatitis B virus activity, CB2 receptor agonists for treatment of chronic pain, aurora kinase inhibitors, spiroimidazolidinone NPC1L1 inhibitors, 1,3,5-Oxadiazinones, selective serotonin 5-HT6 receptor antagonists. It is also used as a pharmaceutical intermediate and as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylpiperidine, is used as a reactant for synthesis of Phenylpropenamide derivatives for anti-hepatitis B virus activity, CB2 receptor agonists for treatment of chronic pain, aurora kinase inhibitors, spiroimidazolidinone NPC1L1 inhibitors, 1,3,5-Oxadiazinones, selective serotonin 5-HT6 receptor antagonists. It is also used as a pharmaceutical intermediate and as a intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-56-2
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00005994
InChI Key JEGMWWXJUXDNJN-UHFFFAOYSA-N
Synonym 3-pipecoline, piperidine, 3-methyl, beta-pipecoline, .beta.-pipecoline, 3-methyl-piperidine, .beta.-methylpiperidine, beta-methylpiperidine, 3-methyl piperidine, 3-pipecoline 8ci, 3ppc
PubChem CID 79081
IUPAC Name 3-methylpiperidine
SMILES CC1CCCNC1

Specifications

Melting Point -24°C
Density 0.845
Boiling Point 124°C to 125°C
Flash Point 8°C (46°F)
Odor Amine-like
Refractive Index 1.447
Quantity 100 g
UN Number UN2733
Beilstein 79807
Solubility Information It is soluble in water.
Formula Weight 99.18
Percent Purity 97%
Chemical Name or Material 3-Methylpiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.