missing translation for 'onlineSavingsMsg'
Learn More

3-Methylphthalic anhydride, 96%, Thermo Scientific Chemicals

Product Code. 11489907
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489907 5 g 5g
11479907 1 g 1g
11499907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489907 Supplier Thermo Scientific Alfa Aesar Supplier No. B22864.06
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

3-Methylphthalic anhydride can react with Methylamine hydrochloride to get 2,3-Dihydro-2,4-dimethyl-1H-isoindole-1,3-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylphthalic anhydride can react with Methylamine hydrochloride to get 2,3-Dihydro-2,4-dimethyl-1H-isoindole-1,3-dione.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under dry inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4792-30-7
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00047316
InChI Key TWWAWPHAOPTQEU-UHFFFAOYSA-N
Synonym 3-methylphthalic anhydride, 4-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-methyl, 3-methyl phthalic anhydride, methylphthalic anhydride, phthalic anhydride, 3-methyl, 2,3-toluenedicarboxylic anhydride, 1,3-isobenzofurandione, methyl, 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione, acmc-1arrb
PubChem CID 98500
IUPAC Name 4-methyl-2-benzofuran-1,3-dione
SMILES CC1=CC=CC2=C1C(=O)OC2=O

Specifications

Melting Point 114°C to 116°C
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 162.14
Percent Purity 96%
Chemical Name or Material 3-Methylphthalic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.