missing translation for 'onlineSavingsMsg'
Learn More

3-methylpentanoic acid, TRC

Product Code. 30474698
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474698 1 g 1g
30470443 500 mg 500mg
30466388 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474698 Supplier Toronto Research Chemicals Supplier No. TRCM2344061G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-43-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
Synonym (±)-3-Methylvaleric acid, Pentanoic acid, 3-methyl- (9CI, ACI), 3-Methylpentanoic acid (ACI), Valeric acid, 3-methyl- (6CI, 7CI, 8CI), Valeric acid, β-methyl- (4CI), 3-Methyl-n-valeric acid, 3-Methylvaleric acid, β-Methylvaleric acid
IUPAC Name 3-methylpentanoic acid
SMILES CCC(C)CC(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H12O2/c1-3-5(2)4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8)
Formula Weight 116.0837
Chemical Name or Material 3-methylpentanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.