missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylnitrosamino)propionitrile, TRC

Product Code. 30136559
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136559 1 g 1g
30134104 250 mg 250mg
17981625 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136559 Supplier Toronto Research Chemicals Supplier No. TRCM3257151G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60153-49-3
Molecular Formula C4 H7 N3 O
Molecular Weight (g/mol) 113.12
Synonym Propionitrile, 3-(methylnitrosamino)- (6CI), 3-(Methylnitrosoamino)propanenitrile, 3-(Methylnitrosamino)propionitrile, 3-(N-Nitrosomethylamino)propionitrile, N-(2-Cyanoethyl)-N-methylnitrosamine
IUPAC Name N-(2-cyanoethyl)-N-methylnitrous amide
SMILES CN(CCC#N)N=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H7N3O/c1-7(6-8)4-2-3-5/h2,4H2,1H3
Formula Weight 113.0589
Chemical Name or Material 3-(Methylnitrosamino)propionitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.