missing translation for 'onlineSavingsMsg'
Learn More

3-Methylisoxazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15299806 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15299806 25 g 25g
15710889 1 g 1g
15289806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15299806 Supplier Thermo Scientific Acros Supplier No. 452660250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4857-42-5
Synonym 3-methylisoxazole-5-carboxylic acid, 3-methyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-methyl, 3-methyl-isoxazole-5-carboxylic acid, 5-carboxy-3-methylisoxazole, 5-isoxazolecarboxylicacid, 3-methyl, 3-methylisoxazole-5-carboxylicacid, acmc-1aruu, 3-methyl-5-carboxyisoxazole, aronis24519
PubChem CID 853085
IUPAC Name 3-methyl-1,2-oxazole-5-carboxylic acid

Specifications

Infrared Spectrum Authentic
Molecular Formula C5H5NO3
Quantity 25 g
InChI Key HXIYCKAAQPHZBM-UHFFFAOYSA-N
SMILES CC1=NOC(=C1)C(=O)O
Molecular Weight (g/mol) 127.099
Formula Weight 127.1
Percent Purity ≥96.0% (HPLC)
Chemical Name or Material 3-Methylisoxazole-5-carboxylic acid
compliance-icons
Product Identifier
  • 3-Methylisoxazole-5-carboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.