missing translation for 'onlineSavingsMsg'
Learn More

3-Methylisoquinoline, 95%

Product Code. 10707918 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10707918 5 g Glass bottle 5g
10156047 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10707918 Supplier Thermo Scientific Acros Supplier No. 428440050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 1125-80-0 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-80-0
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00024139
InChI Key FVVXWRGARUACNW-UHFFFAOYSA-N
Synonym isoquinoline, 3-methyl, 3-methyl-isoquinoline, unii-4quo6nyb5j, 4quo6nyb5j, isoquinolone-3-methyl, 3-methyl-isoquinolin, 3-methyl isoquinoline, 3-methylisoquinoline, 5-20-07-00410 beilstein handbook reference, ksc176i1j
PubChem CID 14306
IUPAC Name 3-methylisoquinoline
SMILES CC1=CC2=CC=CC=C2C=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 63.0°C to 65.0°C
Boiling Point 251.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 3-Methylisoquinoline
compliance-icons
Product Identifier
  • 3-Methylisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.