missing translation for 'onlineSavingsMsg'
Learn More

3-Methylhexahydrophthalic Anhydride, TRC

Product Code. 17913395
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913395 100 mg 100mg
17922595 250 mg 250mg
17933115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17913395 Supplier Toronto Research Chemicals Supplier No. TRCM312385100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57110-29-9
Molecular Formula C9 H12 O3
Molecular Weight (g/mol) 168.19
Synonym Other Names: 1,2-Cyclohexanedicarboxylic anhydride, 3-methyl- (6CI), Hexahydro-4-methyl-1,3-isobenzofurandione, 3-Methyl-1,2-cyclohexanedicarboxylic anhydride, 3-Methylhexahydrophthalic acid anhydride, 3-Methylhexahydrophthalic anhydride, Hexahydro-3-methylphthalic anhydride, Pentadiene-maleic anhydride adduct, 1,2-Cyclohexanedicarboxylic anhydride, 3-methyl- (6CI), Hexahydro-4-methyl-1,3-isobenzofurandione, 3-Methyl-1,2-cyclohexanedicarboxylic anhydride, 3-Methylhexahydrophthalic acid anhydride, 3-Methylhexahydrophthalic anhydride, Hexahydro-3-methylphthalic anhydride, Pentadiene-maleic anhydride adduct
IUPAC Name 4-methyl-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES CC1CCCC2C1C(=O)OC2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H12O3/c1-5-3-2-4-6-7(5)9(11)12-8(6)10/h5-7H,2-4H2,1H3
Formula Weight 168.0786
Chemical Name or Material 3-Methylhexahydrophthalic anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.