missing translation for 'onlineSavingsMsg'
Learn More

3-Methylheptane, 97%

Product Code. 15408949
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408949 1 g 1g
15418949 5 g 5g
15428949 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15408949 Supplier Thermo Scientific Alfa Aesar Supplier No. L12006.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 589-81-1 | C8H18 | 114.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-81-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00027244
InChI Key LAIUFBWHERIJIH-UHFFFAOYSA-N
Synonym heptane, 3-methyl, 2-ethylhexane, 3-methyl-heptane, 2-butylbutane, 3-methyl-heptan, heptane, 3-methyl-, r, acmc-20mdw0, 3-methyl-s-heptane, 3-methylheptane, acmc-1axq2
PubChem CID 11519
IUPAC Name 3-methylheptane
SMILES CCCCC(C)CC

Specifications

Melting Point -121°C
Density 0.71
Boiling Point 118°C to 120°C
Flash Point 7°C (44°F)
Refractive Index 1.398
Quantity 1 g
UN Number UN1262
Beilstein 1730777
Formula Weight 114.23
Percent Purity 97%
Chemical Name or Material 3-Methylheptane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.