missing translation for 'onlineSavingsMsg'
Learn More

3-Methylfuran, 98%

Product Code. 10643532 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10643532 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10643532 Supplier Thermo Scientific Acros Supplier No. 342300010
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 930-27-8 | C5H6O | 82.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 930-27-8
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 65.0°C to 66.0°C
Flash Point −22°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H6O
Refractive Index 1.4310 to 1.4330
MDL Number MFCD00060134
Quantity 1 g
Specific Gravity 0.92
Synonym furan, 3-methyl, 3-methylfurane, 3-methyl furan, 3-methyl-furan, 3-methyl-fura, 3-methylfuran, pubchem19993, 3-methylfuran-methyl-d3, acmc-20an04, 3-methylfuran 1gr
InChI Key KJRRQXYWFQKJIP-UHFFFAOYSA-N
SMILES CC1=COC=C1
IUPAC Name 3-methylfuran
Molecular Weight (g/mol) 82.1
PubChem CID 13587
Formula Weight 82.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylfuran, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methylfuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.