missing translation for 'onlineSavingsMsg'
Learn More

3-Methylenecyclobutanecarbonitrile, 97%

Product Code. 15710869 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15710869 25 g 25g
15700869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15710869 Supplier Thermo Scientific Acros Supplier No. 376010250
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15760-35-7 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15760-35-7
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
InChI Key ZRWMAMOBIQQJSA-UHFFFAOYSA-N
Synonym 3-methylenecyclobutanecarbonitrile, 3-methylenecyclobutane-1-carbonitrile, cyclobutanecarbonitrile, 3-methylene, 3-methylenecyclobutane carbonitrile, 3-methylenecyanocyclobutane, 3-methylene cyclobutyl carbonitrile, 3-methylene-1-cyanocyclobutane, 1-cyano-3-methylidenecyclobutane, pubchem10218, 3-methylenecyclobutanenitrile
PubChem CID 27474
IUPAC Name 3-methylidenecyclobutane-1-carbonitrile
SMILES C=C1CC(C1)C#N

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH2=C4H5CN
Quantity 25 g
Formula Weight 93.13
Percent Purity 97%
Chemical Name or Material 3-Methylenecyclobutanecarbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.