missing translation for 'onlineSavingsMsg'
Learn More

3-Methylenecyclobutanecarbonitrile, 97%, Thermo Scientific™

Product Code. 10324302
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10324302 10 g 10g
10733073 1 g 1g
10533743 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10324302 Supplier Thermo Scientific Maybridge Supplier No. SB02040EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 15760-35-7
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD01085401
InChI Key ZRWMAMOBIQQJSA-UHFFFAOYSA-N
Synonym 3-methylenecyclobutanecarbonitrile, 3-methylenecyclobutane-1-carbonitrile, cyclobutanecarbonitrile, 3-methylene, 3-methylenecyclobutane carbonitrile, 3-methylenecyanocyclobutane, 3-methylene cyclobutyl carbonitrile, 3-methylene-1-cyanocyclobutane, 1-cyano-3-methylidenecyclobutane, pubchem10218, 3-methylenecyclobutanenitrile
PubChem CID 27474
IUPAC Name 3-methylidenecyclobutane-1-carbonitrile
SMILES C=C1CC(C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CH2=C4H5CN
Quantity 10 g
Formula Weight 93.13
Percent Purity 97%
Chemical Name or Material 3-Methylenecyclobutanecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.