missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcytidine, MedChemExpress

Product Code. 16474508
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474508 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16474508 Supplier Medchem Express Supplier No. HY111645
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Methylcytidine, a urinary nucleoside, can be used as a biomarker of four different types of cancer: lung cancer, gastric cancer, colon cancer, and breast cancer.

3-Methylcytidine, a urinary nucleoside, can be used as a biomarker of four different types of cancer: lung cancer, gastric cancer, colon cancer, and breast cancer.

Specifications

Chemical Name or Material 3-Methylcytidine
CAS 2140-64-9
Color White
Purity Grade Notes Research
Molecular Formula C10H15N3O5
Quantity 10 mg
Solubility Information H2O : 90 mg/mL (349.87 mM; Need ultrasonic) ∣DMSO : 83.33 mg/mL (323.94 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C(N(C)C(C=C2)=N)=O)O1)O)O
Molecular Weight (g/mol) 257.24
Formula Weight 257.24
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.