missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcyclohexanone, 97%

Product Code. 10543291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10543291 100 g Glass bottle 100g
10366480 5 g Glass bottle 5g
10459050 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10543291 Supplier Thermo Scientific Acros Supplier No. 126631000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-24-2 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-24-2
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001639
InChI Key UJBOOUHRTQVGRU-UHFFFAOYNA-N
Synonym 3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s
PubChem CID 11567
IUPAC Name 3-methylcyclohexan-1-one
SMILES CC1CCCC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -73.0°C
Color Colorless to Yellow
Density 0.9150g/mL
Boiling Point 169.0°C to 170.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3C6H9(=O)
Refractive Index 1.4440 to 1.4460
Quantity 100 g
Beilstein 07, 15
Specific Gravity 0.915
Solubility Information Solubility in water: insoluble
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methylcyclohexanone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methylcyclohexanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.