missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcatechol, 97%

Product Code. 11489433
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489433 100 g 100g
11479433 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489433 Supplier Thermo Scientific Alfa Aesar Supplier No. A12324.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-17-5 | C7H8O2 | 124.139 g/mol

Used in organic synthesis. Used for the synthesis of antibacterial agent, antioxidants, As a polymer inhibitor, stabilizer, flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in organic synthesis. Used for the synthesis of antibacterial agent, antioxidants, As a polymer inhibitor, stabilizer, flavors.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-17-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00016435
InChI Key PGSWEKYNAOWQDF-UHFFFAOYSA-N
Synonym 3-methylcatechol, 2,3-dihydroxytoluene, 3-methylpyrocatechol, 1,2-benzenediol, 3-methyl, 2,3-toluenediol, dihydroxytoluene, pyrocatechol, 3-methyl, catechol, 3-methyl, 1,2-dihydroxy-3-methylbenzene, 3-methyl-1,2-benzenediol
PubChem CID 340
ChEBI CHEBI:18404
IUPAC Name 3-methylbenzene-1,2-diol
SMILES CC1=C(C(=CC=C1)O)O

Specifications

Melting Point 62°C to 68°C
Boiling Point 241°C
Flash Point 140°C (284°F)
Quantity 100 g
Beilstein 774602
Solubility Information Soluble in water.
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material 3-Methylcatechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.