missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbutyraldehyde, 98%

Product Code. 10606182 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10606182 500 mL Glass bottle 500mL
10114370 100 mL Glass bottle 100mL
10085313 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10606182 Supplier Thermo Scientific Acros Supplier No. 163975000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 590-86-3 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-86-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00007014
InChI Key YGHRJJRRZDOVPD-UHFFFAOYSA-N
Synonym isovaleraldehyde, 3-methylbutyraldehyde, isopentaldehyde, isovaleral, isovalerylaldehyde, isoamylaldehyde, butanal, 3-methyl, isopentanal, beta-methylbutanal, isovaleric aldehyde
PubChem CID 11552
ChEBI CHEBI:16638
IUPAC Name 3-methylbutanal
SMILES CC(C)CC=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -28.0°C
Color Colorless to Yellow
Density 0.7960g/mL
Boiling Point 90.0°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CHO
Refractive Index 1.3870 to 1.3890
Quantity 500 mL
Beilstein 01, 684
Merck Index 15, 5275
Specific Gravity 0.796
Solubility Information Solubility in water: 18.9g/L (25°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.6 mPa.s (20°C)
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylbutyraldehyde, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Methylbutyraldehyde
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.