missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbutanamide, 97%, Thermo Scientific™

Product Code. 10347222
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10347222

Brand: Thermo Scientific Maybridge BTB12988EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 541-46-8
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00014807
InChI Key SANOUVWGPVYVAV-UHFFFAOYSA-N
Synonym isovaleramide, butanamide, 3-methyl, 3-methylbutyramide, isovaleric amide, isopentanamide, beta-methylbutyramide, isovaleric acid amide, unii-9cp4kb634m, isovaleramide usan, .beta.-methylbutyramide
PubChem CID 10930
IUPAC Name 3-methylbutanamide
SMILES CC(C)CC(=O)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material 3-Methylbutanamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.