missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbiphenyl, 95%

Product Code. 11364414
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364414 1 g 1g
11374414 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364414 Supplier Thermo Scientific Alfa Aesar Supplier No. L01006.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 643-93-6 | C13H12 | 168.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643-93-6
Molecular Formula C13H12
Molecular Weight (g/mol) 168.239
MDL Number MFCD00008533
InChI Key NPDIDUXTRAITDE-UHFFFAOYSA-N
Synonym 3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
PubChem CID 12564
IUPAC Name 1-methyl-3-phenylbenzene
SMILES CC1=CC=CC(=C1)C2=CC=CC=C2

Specifications

Melting Point 4°C to 5°C
Density 1.014
Boiling Point 272°C
Flash Point >110°C (230°F)
Refractive Index 1.602
Quantity 1 g
Beilstein 2039658
Formula Weight 168.24
Percent Purity 95%
Chemical Name or Material 3-Methylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.