missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzylamine, 98%

Product Code. 11311767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311767 5 g 5g
11301767 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311767 Supplier Thermo Scientific Alfa Aesar Supplier No. L04922.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 100-81-2 | C8H11N | 121.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-81-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008118
InChI Key RGXUCUWVGKLACF-UHFFFAOYSA-N
Synonym 3-methylbenzylamine, m-tolylmethanamine, 3-methylphenyl methanamine, benzenemethanamine, 3-methyl, m-methylbenzylamine, 1-3-methylphenyl methanamine, 3-methylbenzyl amine, alpha-amino-m-xylene, benzylamine, m-methyl, m-xylylamine
PubChem CID 66015
IUPAC Name (3-methylphenyl)methanamine
SMILES CC1=CC(=CC=C1)CN

Specifications

Density 0.96
Boiling Point 203°C to 205°C
Flash Point 80°C (176°F)
Refractive Index 1.536
Quantity 5 g
UN Number UN2735
Beilstein 507474
Sensitivity Air Sensitive
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 3-Methylbenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.