missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzylamine, 98%

Product Code. 10205920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205920 5 g Glass bottle 5g
10051721 1 g Glass bottle 1g
10429260 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10205920 Supplier Thermo Scientific Acros Supplier No. 155480050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-81-2 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-81-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008118
InChI Key RGXUCUWVGKLACF-UHFFFAOYSA-N
Synonym 3-methylbenzylamine, m-tolylmethanamine, 3-methylphenyl methanamine, benzenemethanamine, 3-methyl, m-methylbenzylamine, 1-3-methylphenyl methanamine, 3-methylbenzyl amine, alpha-amino-m-xylene, benzylamine, m-methyl, m-xylylamine
PubChem CID 66015
IUPAC Name (3-methylphenyl)methanamine
SMILES CC1=CC(=CC=C1)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9660g/mL
Boiling Point 202.0°C to 205.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2NH2
Refractive Index 1.5350 to 1.5370
Quantity 5 g
Beilstein 12, 1134
Specific Gravity 0.966
Formula Weight 121.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylbenzylamine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.