missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzyl chloride, 98%

Product Code. 11489692
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489692 500 g 500g
11469692 25 g 25g
11479692 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489692 Supplier Thermo Scientific Alfa Aesar Supplier No. A10784.36
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 620-19-9 | C8H9Cl | 140.61 g/mol

3-Methylbenzyl chloride is used as an organic light-emitting diode and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylbenzyl chloride is used as an organic light-emitting diode and as an intermediate in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-19-9
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000909
InChI Key LZBOHNCMCCSTJX-UHFFFAOYSA-N
Synonym 3-methylbenzyl chloride, 1-chloromethyl-3-methylbenzene, m-xylyl chloride, 3-methylbenzylchloride, alpha-chloro-m-xylene, m-methylbenzyl chloride, 1-chloromethyl-3-methyl-benzene, m-chloromethyl toluene, 3-chloromethyl toluene, benzene, 1-chloromethyl-3-methyl
PubChem CID 12102
IUPAC Name 1-(chloromethyl)-3-methylbenzene
SMILES CC1=CC(=CC=C1)CCl

Specifications

Density 1.066
Boiling Point 195°C to 196°C
Flash Point 75°C (167°F)
Odor Pungent
Refractive Index 1.535
Quantity 500 g
UN Number UN3265
Beilstein 2040357
Merck Index 14,10091
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 140.61
Percent Purity 98%
Chemical Name or Material 3-Methylbenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.