missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzyl bromide, 97%

Product Code. 11436426
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436426 25 g 25g
11426426 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436426 Supplier Thermo Scientific Alfa Aesar Supplier No. B24731.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 620-13-3 | C8H9Br | 185.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-13-3
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000177
InChI Key FWLWTILKTABGKQ-UHFFFAOYSA-N
Synonym 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo
PubChem CID 12099
IUPAC Name 1-(bromomethyl)-3-methylbenzene
SMILES CC1=CC(=CC=C1)CBr

Specifications

Density 1.37
Boiling Point 184°C to 185°C (340 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.569
Quantity 25 g
UN Number UN1701
Beilstein 2076425
Merck Index 14,10090
Formula Weight 185.07
Percent Purity 97%
Chemical Name or Material 3-Methylbenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.