missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzophenone, 98+%

Product Code. 11404137
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404137 10 g 10g
11414137 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404137 Supplier Thermo Scientific Alfa Aesar Supplier No. A19568.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 643-65-2 | C14H12O | 196.25 g/mol

3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 643-65-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00008535
InChI Key URBLVRAVOIVZFJ-UHFFFAOYSA-N
Synonym 3-methylbenzophenone, 3-methyl-benzophenone, phenyl m-tolyl methanone, benzophenone, 3-methyl, 3-methylphenyl-phenylmethanone, methanone, 3-methylphenyl phenyl, 3-methylphenyl phenyl ketone, 3-benzoyltoluene, m-methylbenzophenone, zlchem 905
PubChem CID 69511
IUPAC Name (3-methylphenyl)-phenylmethanone
SMILES CC1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Specifica

Density 1.093
Boiling Point 316°C
Flash Point >110°C (230°F)
Refractive Index 1.598
Quantity 10 g
Beilstein 2045488
Solubility Information Difficult to mix.
Formula Weight 196.25
Percent Purity 99%
Chemical Name or Material 3-Methylbenzophenone

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.