missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzofuran, 98%

Product Code. 10190502 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Glass bottle
missing translation for 'unitSize'
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10190502 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10190502 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 358530010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21535-97-7 | C9H8O | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21535-97-7
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H8O
Refractive Index 1.5535 to 1.5555
MDL Number MFCD00205641
Quantity 1 g
Synonym 3-methylbenzofuran, 3-methyl-benzofuran, benzofuran, 3-methyl, 3-methylbenzo b furan, acmc-209flj, #
InChI Key ZRXHLJNBNWVNIM-UHFFFAOYSA-N
SMILES CC1=COC2=CC=CC=C12
IUPAC Name 3-methyl-1-benzofuran
Molecular Weight (g/mol) 132.16
PubChem CID 88939
Formula Weight 132.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylbenzofuran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.