missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzo[b]thiophene-2-carboxaldehyde, 98%

Product Code. 11400193
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400193 25 g 25g
11480183 1 g 1g
11490183 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400193 Supplier Thermo Scientific Alfa Aesar Supplier No. A11375.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22053-74-3 | C10H8OS | 176.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22053-74-3
Molecular Formula C10H8OS
Molecular Weight (g/mol) 176.233
MDL Number MFCD00052307
InChI Key DRZGHNXLEQHVHB-UHFFFAOYSA-N
Synonym 3-methylbenzo b thiophene-2-carboxaldehyde, 3-methylbenzo b thiophene-2-carbaldehyde, 3-methyl-benzo b thiophene-2-carbaldehyde, pubchem7453, acmc-20an0e, 3-methylthianaphthene-2-carboxaldehyde, 3-methyl-2-benzothiophenecarboxaldehyde, 3-methyl benzo b thiophene-2-carbaldehyde, 3-methylbenzo b thiophene-2 carboxaldehyde
PubChem CID 519918
IUPAC Name 3-methyl-1-benzothiophene-2-carbaldehyde
SMILES CC1=C(SC2=CC=CC=C12)C=O

Specifications

Melting Point 88°C to 90°C
Quantity 25 g
Beilstein 121582
Sensitivity Air Sensitive
Formula Weight 176.24
Percent Purity 98%
Chemical Name or Material 3-Methylbenzo[b]thiophene-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.