missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzenethiol, 95%

Product Code. 10280060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10280060 1 g Glass bottle 1g
11391056 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10280060 Supplier Thermo Scientific Acros Supplier No. 421270010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 108-40-7 | C7H8S | 124.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-40-7
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004843
InChI Key WRXOZRLZDJAYDR-UHFFFAOYSA-N
Synonym m-thiocresol, m-toluenethiol, 3-methylthiophenol, 3-thiocresol, benzenethiol, 3-methyl, m-methylbenzenethiol, m-methylthiophenol, 3-mercaptotoluene, m-tolylmercaptan, m-mercaptotoluene
PubChem CID 7930
IUPAC Name 3-methylbenzene-1-thiol
SMILES CC1=CC=CC(S)=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0440g/mL
Boiling Point 77.0°C (10.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SH
Refractive Index 1.5742 to 1.5772
Quantity 1 g
Merck Index 15,9478
Specific Gravity 1.044
Formula Weight 124.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Methylbenzenethiol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • m-Toluenethiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.