missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzenesulfonic Acid Monohydrate, TRC

Product Code. 30484239
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484239 100 mg 100mg
30480835 1 g 1g
30470666 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484239 Supplier Toronto Research Chemicals Supplier No. TRCM276595100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 312619-56-0
Molecular Formula C7 H8 O3 S . H2 O
Molecular Weight (g/mol) 190.22
Synonym Benzenesulfonic acid, 3-methyl-, hydrate (1:1), Benzenesulfonic acid, 3-methyl-, hydrate (1:1) (ACI), Benzenesulfonic acid, 3-methyl-, monohydrate (9CI), m-Toluenesulfonic Acid Monohydrate, GW845032A, 3-Methylbenzenesulfonic acid hydrate, m-TSA, GSK3985771C (Niraparib related)
IUPAC Name 3-methylbenzenesulfonic acid;hydrate
SMILES O.Cc1cccc(c1)S(=O)(=O)O

Specifica

Quantity 100 mg
InChI Formula InChI=1S/C7H8O3S.H2O/c1-6-3-2-4-7(5-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2
Formula Weight 190.023
Chemical Name or Material 3-Methylbenzenesulfonic Acid Monohydrate
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.