missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzene-1,2-diamine, 97%, Thermo Scientific™

Product Code. 10306894
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306894 10 g Amber Glass Bottle 10g
10662265 1 g Amber Glass Bottle 1g
10366534 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10306894 Supplier Thermo Scientific Maybridge Supplier No. BTB07447EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2687-25-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00011589
InChI Key AXNUJYHFQHQZBE-UHFFFAOYSA-N
Synonym 2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
PubChem CID 17593
IUPAC Name 3-methylbenzene-1,2-diamine
SMILES CC1=CC=CC(N)=C1N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3C6H3(NH2)2
Quantity 10 g
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 3-Methylbenzene-1, 2-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.