missing translation for 'onlineSavingsMsg'
Learn More

3-Methylamino-1-propanol, 95%

Product Code. 15455018
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455018 5 g 5g
15445018 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455018 Supplier Thermo Scientific Alfa Aesar Supplier No. H58818.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42055-15-2 | C4H11NO | 89.138 g/mol

3-Methylamino-1-propanol is an alkanolamine that is used to redisperse compacted solids (such as antiperspirant materials) in order to analyze its components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylamino-1-propanol is an alkanolamine that is used to redisperse compacted solids (such as antiperspirant materials) in order to analyze its components.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42055-15-2
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD06797456
InChI Key KRGXWTOLFOPIKV-UHFFFAOYSA-N
Synonym 3-methylamino propan-1-ol, 3-methylamino-1-propanol, 3-methylaminopropanol, 3-methylamino-propanol, 3-methylamino propanol, acmc-1argo, 3-methylamino-propan-1-ol, 3-hydroxypropyl methylamine, n-methyl-3-hydroxypropylamine
PubChem CID 10148986
IUPAC Name 3-(methylamino)propan-1-ol
SMILES CNCCCO

Specifications

Density 0.98
Boiling Point 169°C
Flash Point 82°C (180°F)
Linear Formula CH3NH(CH2)3OH
Refractive Index 1.44
Quantity 5 g
UN Number UN2735
Solubility Information Slightly soluble in water.
Formula Weight 89.14
Percent Purity 95%
Chemical Name or Material 3-Methylamino-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.