missing translation for 'onlineSavingsMsg'
Learn More

3-Methylamino-1,2-propanediol, 99%

Product Code. 11333296
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333296 100 g 100g
11323296 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333296 Supplier Thermo Scientific Alfa Aesar Supplier No. L12653.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40137-22-2 | C4H11NO2 | 105.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40137-22-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00082732
InChI Key WOMTYMDHLQTCHY-UHFFFAOYSA-N
Synonym 3-methylamino-1,2-propanediol, 3-methylamino propane-1,2-diol, 1,2-propanediol, 3-methylamino, acmc-209jb1, ksc495i3p, 3-methylamino-1,2-propandiol, n-methylamino-2,3-propanediol, 3-methylamino-propan-1,2-diol, 3-methylamino-propane-1,2-diol
PubChem CID 2734122
IUPAC Name 3-(methylamino)propane-1,2-diol
SMILES CNCC(CO)O

Specifications

Density 1.093
Boiling Point 239°C to 241°C
Flash Point 152°C (305°F)
Refractive Index 1.4745
Quantity 100 g
UN Number UN2735
Beilstein 1733341
Formula Weight 105.14
Percent Purity 99%
Chemical Name or Material 3-Methylamino-1,2-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.