missing translation for 'onlineSavingsMsg'
Learn More

3-Methyladenine, MedChemExpress

Product Code. 16403697
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16403697

Brand: Medchem Express HY19312

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Methyladenine (3-MA) is a PI3K inhibitor. 3-Methyladenine is a widely used inhibitor of autophagy via its inhibitory effect on class III PI3K.

3-Methyladenine (3-MA) is a PI3K inhibitor. 3-Methyladenine is a widely used inhibitor of autophagy via its inhibitory effect on class III PI3K.

Specifications

Chemical Name or Material 3-Methyladenine
Physical Form Solid
CAS 5142-23-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H7N5
For Use With (Application) Cancer-Kinase/protease
Synonym 3-MA
Solubility Information H2O : 5 mg/mL (33.52 mM; Need ultrasonic)
SMILES NC1=C2N=CN=C2N(C)C=N1
Molecular Weight (g/mol) 149.15
Formula Weight 149.15
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.