missing translation for 'onlineSavingsMsg'
Learn More

3-Methyladenine, MedChemExpress

Product Code. 16491896
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491896 500 mg 500mg
16461286 200 mg 200mg
16427586 100 mg 100mg
16403697 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491896 Supplier Medchem Express Supplier No. HY19312
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Methyladenine (3-MA) is a PI3K inhibitor. 3-Methyladenine is a widely used inhibitor of autophagy via its inhibitory effect on class III PI3K.

3-Methyladenine (3-MA) is a PI3K inhibitor. 3-Methyladenine is a widely used inhibitor of autophagy via its inhibitory effect on class III PI3K.

Specifications

Chemical Name or Material 3-Methyladenine
Physical Form Solid
CAS 5142-23-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C6H7N5
For Use With (Application) Cancer-Kinase/protease
Synonym 3-MA
Solubility Information H2O : 5 mg/mL (33.52 mM; Need ultrasonic)
SMILES NC1=C2N=CN=C2N(C)C=N1
Molecular Weight (g/mol) 149.15
Formula Weight 149.15
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.