missing translation for 'onlineSavingsMsg'
Learn More

3'-Methylacetophenone, 97%

Product Code. 10057939 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10057939 25 g Glass bottle 25g
10119399 5 g Glass bottle 5g
10017989 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10057939 Supplier Thermo Scientific Acros Supplier No. 126170250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 585-74-0 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-74-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008742
InChI Key FSPSELPMWGWDRY-UHFFFAOYSA-N
Synonym 3'-methylacetophenone, 1-m-tolyl ethanone, 3-methylacetophenone, m-methylacetophenone, ethanone, 1-3-methylphenyl, acetophenone, m-methyl, methyl m-tolyl ketone, 1-3-methylphenyl ethan-1-one, 3-acetyltoluene, acetophenone, 3'-methyl
PubChem CID 11455
IUPAC Name 1-(3-methylphenyl)ethanone
SMILES CC(=O)C1=CC=CC(C)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -9.0°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 218.0°C to 220.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4COCH3
Refractive Index 1.529
Quantity 25 g
Beilstein 07,307
Specific Gravity 1.01
Formula Weight 134.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3′-Methylacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.