missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-N-methylbenzylamine, 97%, Thermo Scientific™

Product Code. p-7041660
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15439818

Brand: Thermo Scientific Alfa Aesar H63231.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Methyl-N-methylbenzylamine, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39180-84-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD05865181
InChI Key WCTNVGNEUDTSOZ-UHFFFAOYSA-N
Synonym 3-methyl-n-methylbenzylamine, n-methyl-1-m-tolyl methanamine, 3-methylbenzyl methylamine, methyl 3-methylphenyl methyl amine, benzenemethanamine, n,3-dimethyl, acmc-20a7fx, n,3-dimethylbenzylamine, methyl 3-methylbenzyl amine, n-methyl-3-methylbenzylamine, ksc493m5b
PubChem CID 13794465
IUPAC Name N-methyl-1-(3-methylphenyl)methanamine
SMILES CC1=CC(=CC=C1)CNC

Specifications

Density 0.9
Boiling Point 90°C to 92°C (15 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.519
Quantity 1 g
UN Number UN3259
Solubility Information Slightly soluble in water.
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 3-Methyl-N-methylbenzylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.