missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-L-phenylalanine, 95%, Thermo Scientific Chemicals
£26.80 - £321.00
Chemical Identifiers
CAS | 114926-37-3 |
---|---|
Molecular Formula | C10H13NO2 |
Molecular Weight (g/mol) | 179.219 |
MDL Number | MFCD00044972 |
InChI Key | JZRBSTONIYRNRI-VIFPVBQESA-N |
Synonym | 3-methyl-l-phenylalanine, l-3-methylphenylalanine, 3-methylphenyl-l-alanine, 3-methy-l-phenylalanine, s-2-amino-3-m-tolyl propanoic acid, 3-methylphenylalanine, l-3-methylphe, phenylalanine, 3-methyl, 2s-2-amino-3-3-methylphenyl propanoic acid, l-3-me-phe-oh |
PubChem CID | 2761494 |
IUPAC Name | (2S)-2-amino-3-(3-methylphenyl)propanoic acid |
SMILES | CC1=CC(=CC=C1)CC(C(=O)O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468537
|
Thermo Scientific Alfa Aesar
H52047.MD |
250 mg |
£26.80
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15448537
|
Thermo Scientific Alfa Aesar
H52047.03 |
1 g |
£75.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15458537
|
Thermo Scientific Alfa Aesar
H52047.06 |
5 g |
£321.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application in the identification of N-acyl 4-(5-pyrimidine-2,4-dionyl)phenylalanine derivatives and their orally active prodrug esters as dual-acting alpha4-beta1 and alpha4-beta7 receptor antagonists.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
114926-37-3 | |
179.219 | |
JZRBSTONIYRNRI-VIFPVBQESA-N | |
2761494 | |
CC1=CC(=CC=C1)CC(C(=O)O)N |
C10H13NO2 | |
MFCD00044972 | |
3-methyl-l-phenylalanine, l-3-methylphenylalanine, 3-methylphenyl-l-alanine, 3-methy-l-phenylalanine, s-2-amino-3-m-tolyl propanoic acid, 3-methylphenylalanine, l-3-methylphe, phenylalanine, 3-methyl, 2s-2-amino-3-3-methylphenyl propanoic acid, l-3-me-phe-oh | |
(2S)-2-amino-3-(3-methylphenyl)propanoic acid |
Specifications
114926-37-3 | |
MFCD00044972 | |
Slightly soluble in water. | |
CC1=CC(=CC=C1)CC(C(=O)O)N | |
179.219 | |
179.22 | |
3-Methyl-L-phenylalanine |
C10H13NO2 | |
3-methyl-l-phenylalanine, l-3-methylphenylalanine, 3-methylphenyl-l-alanine, 3-methy-l-phenylalanine, s-2-amino-3-m-tolyl propanoic acid, 3-methylphenylalanine, l-3-methylphe, phenylalanine, 3-methyl, 2s-2-amino-3-3-methylphenyl propanoic acid, l-3-me-phe-oh | |
JZRBSTONIYRNRI-VIFPVBQESA-N | |
(2S)-2-amino-3-(3-methylphenyl)propanoic acid | |
2761494 | |
95% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only