missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl Hippuric Acid, TRC

Product Code. 30137376
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137376 5 mg 5mg
30135679 100 mg 100mg
17982815 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137376 Supplier Toronto Research Chemicals Supplier No. TRCM3119855MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27115-49-7
Molecular Formula C10 H11 N O3
Molecular Weight (g/mol) 193.2
Synonym Glycine, N-(3-methylbenzoyl)-, Hippuric acid, 3-methyl- (7CI), Hippuric acid, m-methyl- (6CI,8CI), N-(3-Methylbenzoyl)glycine, (3-Methyl-benzoylamino)-acetic acid, 2-[(3-Methylbenzoyl)amino]acetic acid, 2-[(3-Methylphenyl)formamido]acetic acid, 3-Methylbenzoylglycine, 3-Methylhippuric acid, NSC 201735, m-Methylhippuric acid, m-Toluric acid
IUPAC Name 2-[(3-methylbenzoyl)amino]acetic acid
SMILES Cc1cccc(c1)C(=O)NCC(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H11NO3/c1-7-3-2-4-8(5-7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)
Formula Weight 193.0739
Chemical Name or Material 3-Methyl Hippuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.