missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl Adenine, TRC

Product Code. 30468082
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468082 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468082 Supplier Toronto Research Chemicals Supplier No. TRCM27590425MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methyl Adenine
CAS 5142-23-4
Molecular Formula C6 H7 N5
Quantity 25 mg
Synonym 3-Methyl-3H-purin-6-amine, 3-Methyl-6-aminopurine, 3-Methyladenine, 6-Amino-3-methylpurine, N3-Methyladenine, NSC 66389
InChI Formula InChI=1S/C6H7N5/c1-11-3-10-5(7)4-6(11)9-2-8-4/h2-3H,7H2,1H3
SMILES Cn1cnc(N)c2ncnc12
IUPAC Name 3-methylpurin-6-amine
Molecular Weight (g/mol) 149.15
Formula Weight 149.0701
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.