missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethyl)pyrazole, 99%, Thermo Scientific™

Product Code. 10766285 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10766285 1 g Glass bottle 1g
10235104 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10766285 Supplier Thermo Scientific Acros Supplier No. 398600010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10010-93-2
Molecular Formula C5H5F3N2
Molecular Weight (g/mol) 150.1
MDL Number MFCD00829309
InChI Key DLCHCAYDSKIFIN-UHFFFAOYSA-N
Synonym 5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
PubChem CID 139077
IUPAC Name 5-methyl-3-(trifluoromethyl)-1H-pyrazole
SMILES CC1=CC(=NN1)C(F)(F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 88.0°C to 90.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 150.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)pyrazole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-5-(trifluoromethyl)pyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.