missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethoxy)benzamide, 97%, Thermo Scientific™

Product Code. 15474257
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474257 5 g 5g
15464257 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474257 Supplier Thermo Scientific Alfa Aesar Supplier No. H31653.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-53-6
Molecular Formula C9H8F3NO2
Molecular Weight (g/mol) 219.163
MDL Number MFCD09025372
InChI Key QKDMEDIWAQNUHI-UHFFFAOYSA-N
Synonym 3-methyl-5-trifluoromethoxy benzamide, 5-methyl-3-trifluoromethoxy benzamide
PubChem CID 46737539
IUPAC Name 3-methyl-5-(trifluoromethoxy)benzamide
SMILES CC1=CC(=CC(=C1)C(=O)N)OC(F)(F)F

Specifikationer

Melting Point 148°C to 152°C
Quantity 5 g
Formula Weight 219.16
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(trifluoromethoxy)benzamide
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.